Assay Details
| Gene Symbol: Prcp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106281.2 | NP_001099751.2 | 1 - 2 | 222 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of thyroid hormone mediated signaling pathway
plasma kallikrein-kinin cascade
negative regulation of systemic arterial blood pressure
proteolysis
biological_process
glucose homeostasis
regulation of blood vessel endothelial cell migration
angiogenesis involved in wound healing
energy homeostasis
regulation of reactive oxygen species metabolic process
molecular_function
serine-type peptidase activity
regulation of thyroid hormone mediated signaling pathway
plasma kallikrein-kinin cascade
negative regulation of systemic arterial blood pressure
proteolysis
biological_process
glucose homeostasis
regulation of blood vessel endothelial cell migration
angiogenesis involved in wound healing
energy homeostasis
regulation of reactive oxygen species metabolic process
molecular_function
serine-type peptidase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P113184 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 327908 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 971703 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 125864 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 369682 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 958197 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 421080 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412439 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26406 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 72669 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 91476 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206619 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 900968 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 502597 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 748111 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337908 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744967 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 502969 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 525997 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569435 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121197 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249667 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106992 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 784455 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 757609 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 604985 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 23317 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685036 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221888 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203745 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546532 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897362 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607807 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 242608 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 748941 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 25924 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63866 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162523 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75800 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 96382 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10909 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 85152 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 67174 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P113184 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P113184 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P113184 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P20547 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P682439 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 44551 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10708680 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708674 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708677 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708681 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708678 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708676 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708675 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708679 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708673 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10708672 | - | - |
| Rat Genome 230 2.0 Array | 1393657_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1379248_at | - | - |
| Rat Genome 230 2.0 Array | 1393657_at | - | - |
| Rat Genome 230 2.0 Array | 1380279_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1380279_at | - | - |
| Rat Genome 230 2.0 Array | 1379248_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1379248_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1379248_at | - | - |
| Affymetrix HT Rat Focus Array | 1393657_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1393657_at | - | - |
| Affymetrix HT Rat Focus Array | 1380279_at | - | - |
| Affymetrix HT Rat Focus Array | 1380279_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100015734.rn.1 | TC0100002044.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100015733.rn.1 | TC0100002044.rn.1 | RTA TC |
| Rat Expression Set 230 | 1393657_at | - | - |
| Rat Expression Set 230 | 1393657_PM_at | - | - |
| Rat Expression Set 230 | 1379248_at | - | - |
| Rat Expression Set 230 | 1380279_PM_at | - | - |
| Rat Expression Set 230 | 1380279_at | - | - |
| Rat Expression Set 230 | 1379248_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708672 | - | - |
| Rat Genome U34 Set | rc_AI101363_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21193128 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1365752 | - | - |

Prcp
Genomic Map