Assay Details
| Gene Symbol: rnf141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001800.1 | NP_001001800.1 | 4 - 5 | 544 | 79 | |
| XM_006230050.2 | XP_006230112.1 | 4 - 5 | 564 | 79 | ||
| XM_008759730.1 | XP_008757952.1 | 5 - 6 | 670 | 79 | ||
| XM_008759731.1 | XP_008757953.1 | 4 - 5 | 537 | 79 | ||
| GenBank mRNA | AY621087.1 | - | 4 - 5 | 544 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

rnf141
Genomic Map