Assay Details
| Gene Symbol: Calca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017338.2 | NP_059034.1 | 3 - 4 | 336 | 75 | |
| XM_008759676.1 | XP_008757898.1 | 3 - 4 | 387 | 75 | ||
| GenBank mRNA | M26137.1 | - | 2 - 3 | 241 | 75 | |
| V01228.1 | - | 3 - 4 | 297 | 75 | ||
| V01229.1 | - | 2 - 3 | 241 | 75 | ||
More Information
Set Membership: |
|

Calca
Genomic Map