Assay Details
Gene Symbol: Zfp318 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008766902.1 | XP_008765124.1 | 4 - 5 | 731 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
