Assay Details
| Gene Symbol: Mrpl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034136.1 | NP_001029308.1 | 3 - 4 | 434 | 78 | |
| GenBank mRNA | BC100081.1 | - | 3 - 4 | 434 | 7325700 | 78 |
| FQ214924.1 | - | 3 - 4 | 467 | 78 | ||
| FQ229593.1 | - | 3 - 4 | 468 | 78 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mrpl2
Genomic Map