Assay Details
| Gene Symbol: Smap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750667.3 | XP_003750715.3 | 6 - 7 | 813 | 67 | |
| XM_003750668.3 | XP_003750716.3 | 7 - 8 | 895 | 67 | ||
More Information
Set Membership: |
|

Smap1
Genomic Map