Assay Details
| Gene Symbol: Mut | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001067239.5 | XP_001067239.3 | 8 - 9 | 1814 | 73 | |
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P586025 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P586025 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P100823 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P100823 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P586025 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P586025 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P100823 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10926793 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10922009 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10922003 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926790 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926788 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10922006 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10922012 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926792 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926786 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10922010 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926783 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926789 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10922005 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10922004 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926795 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10922015 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10922008 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926785 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926791 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926794 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10922013 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10922007 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926784 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10922014 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10922011 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926787 | - | - |
| Rat Genome 230 2.0 Array | 1393152_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1393152_at | - | - |
| Rat Genome 230 2.0 Array | 1385690_at | - | - |
| Rat Genome 230 2.0 Array | 1385690_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1382748_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1382748_at | - | - |
| Affymetrix HT Rat Focus Array | 1382748_at | - | - |
| Affymetrix HT Rat Focus Array | 1382748_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1385690_at | - | - |
| Affymetrix HT Rat Focus Array | 1385690_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1393152_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1393152_at | - | - |
| Rat Expression Set 230 | 1393152_PM_at | - | - |
| Rat Expression Set 230 | 1382748_at | - | - |
| Rat Expression Set 230 | 1393152_at | - | - |
| Rat Expression Set 230 | 1385690_PM_at | - | - |
| Rat Expression Set 230 | 1385690_at | - | - |
| Rat Expression Set 230 | 1382748_PM_at | - | - |
| Rat Genome U34 Set | rc_AI007821_at | - | - |
| Rat Genome U34 Set | rc_AA848752_at | - | - |

Mut
Genomic Map