Assay Details
| Gene Symbol: Slc35b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037215.3 | NP_001032292.1 | 1 - 2 | 123 | 52 | |
| GenBank mRNA | AB197928.1 | - | 1 - 2 | 73 | 52 | |
| BC100647.1 | - | 1 - 2 | 123 | 7932661 | 52 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AB197928.1,BC100647.1,NM_001037215.3 transcripts.
This assay is in the 5'UTR region of the AB197928.1,BC100647.1,NM_001037215.3 transcripts.
Set Membership: |
|

Slc35b2
Genomic Map