Assay Details
| Gene Symbol: Ckmt1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012738.1 | NP_001012756.1 | 3 - 4 | 648 | 78 | |
| XM_006234830.2 | XP_006234892.1 | 4 - 5 | 604 | 78 | ||
| GenBank mRNA | BC091335.1 | - | 3 - 4 | 648 | 7314893 | 78 |
| X59737.1 | - | 4 - 5 | 558 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ckmt1b
Genomic Map