Assay Details
| Gene Symbol: Ube2e3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047857.3 | NP_001041322.2 | 3 - 4 | 626 | 100 | |
| XM_006234405.2 | XP_006234467.1 | 3 - 4 | 553 | 100 | ||
| GenBank mRNA | DQ480743.1 | - | 3 - 4 | 629 | 100 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ube2e3
Genomic Map