Assay Details
| Gene Symbol: Caprin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012185.2 | NP_001012185.2 | 7 - 8 | 903 | 72 | |
| XM_006234652.2 | XP_006234714.1 | 8 - 9 | 958 | 72 | ||
| GenBank mRNA | AB373991.1 | - | 6 - 7 | 818 | 72 | |
| AY155572.1 | - | 6 - 7 | 759 | 72 | ||
| BC087123.1 | - | 6 - 7 | 773 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P042117 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P817539 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536040 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534952 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 410353 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 568857 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251825 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628907 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 844347 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966065 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 289681 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586459 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 651539 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 850191 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 67324 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304124 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 441424 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 697206 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379708 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 812205 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 822972 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88578 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 954442 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70141 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 173918 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 51166 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P817539 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P042117 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P817539 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P042117 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P042117 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P817539 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1027438 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P10962 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P817539 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 64071 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 58989 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 4071 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10847860 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10847868 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10847861 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10847866 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10847869 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10847865 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10847864 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10847858 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10847856 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10847862 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10847863 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10847857 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10847855 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10847867 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10847854 | - | - |
| Rat Genome 230 2.0 Array | 1395412_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1395173_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1395173_at | - | - |
| Rat Genome 230 2.0 Array | 1395412_at | - | - |
| Rat Genome 230 2.0 Array | 1371938_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1371938_at | - | - |
| Rat Genome 230 2.0 Array | 1371939_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1371939_at | - | - |
| Affymetrix HT Rat Focus Array | 1395173_at | - | - |
| Affymetrix HT Rat Focus Array | 1395173_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1371939_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1371939_at | - | - |
| Affymetrix HT Rat Focus Array | 1380358_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1380358_at | - | - |
| Affymetrix HT Rat Focus Array | 1395412_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1395412_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0300028757.rn.1 | TC0300003773.rn.1 | RTA TC |
| Rat Expression Set 230 | 1371939_PM_at | - | - |
| Rat Expression Set 230 | 1371938_at | - | - |
| Rat Expression Set 230 | 1371939_at | - | - |
| Rat Expression Set 230 | 1371938_PM_at | - | - |
| Rat Expression Set 230 | 1395173_PM_at | - | - |
| Rat Expression Set 230 | 1395173_at | - | - |
| Rat Expression Set 230 | 1395412_PM_at | - | - |
| Rat Expression Set 230 | 1395412_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10847854 | - | - |
| Rat Genome U34 Set | rc_AI008787_at | - | - |
| Rat Genome U34 Set | rc_AI171369_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21922542 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376498 | - | - |

Caprin1
Genomic Map