Assay Details
| Gene Symbol: Klhl41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057191.1 | NP_476539.1 | 1 - 2 | 1110 | 71 | |
| GenBank mRNA | AJ293948.1 | - | 1 - 2 | 1110 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Klhl41
Genomic Map