Assay Details
| Gene Symbol: Ppp1r14d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172011.1 | NP_742008.1 | 1 - 2 | 297 | 66 | |
| GenBank mRNA | AY122322.1 | - | 1 - 2 | 297 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppp1r14d
Genomic Map