Assay Details
Gene Symbol: Tmx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100529.1 | NP_001093999.1 | 6 - 7 | 753 | 71 | |
GenBank mRNA | BC094304.1 | - | 3 - 4 | 185 | 71 | |
BC109379.1 | - | 3 - 4 | 185 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
