Assay Details
| Gene Symbol: Rbms1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012184.1 | NP_001012184.1 | 11 - 12 | 1493 | 62 | |
| XM_006234234.2 | XP_006234296.1 | 12 - 13 | 1298 | 62 | ||
| XM_006234235.2 | XP_006234297.1 | 12 - 13 | 1591 | 62 | ||
| XM_006234236.2 | XP_006234298.1 | 12 - 13 | 1390 | 62 | ||
| XM_006234237.2 | XP_006234299.1 | 12 - 13 | 2150 | 62 | ||
| XM_006234238.2 | XP_006234300.1 | 11 - 12 | 1250 | 62 | ||
| XM_006234239.2 | XP_006234301.1 | 13 - 14 | 1788 | 62 | ||
| XM_006234241.2 | XP_006234303.1 | 11 - 12 | 1340 | 62 | ||
| XM_008761860.1 | XP_008760082.1 | 12 - 13 | 1879 | 62 | ||
| GenBank mRNA | BC087094.1 | - | 11 - 12 | 1493 | 7191334 | 62 |
More Information
Updated Mapping Information
Set Membership: |
|

Rbms1
Genomic Map