Assay Details
Gene Symbol: LOC691952 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109652.1 | NP_001103122.1 | 1 - 2 | 84 | 69 | |
XM_008762201.1 | XP_008760423.1 | 1 - 2 | 286 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
