Assay Details
Gene Symbol: LOC691952 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109652.1 | NP_001103122.1 | 4 - 5 | 370 | 131 | |
XM_008762201.1 | XP_008760423.1 | 4 - 5 | 572 | 131 |

More Information
Set Membership: |
![]() ![]() ![]() |
