Assay Details
| Gene Symbol: Stard7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106503.1 | NP_001099973.1 | 3 - 4 | 838 | 80 | |
| XM_006234917.2 | XP_006234979.1 | 5 - 6 | 1096 | 80 | ||
| XM_008762156.1 | XP_008760378.1 | 5 - 6 | 1100 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stard7
Genomic Map