Assay Details
| Gene Symbol: Zfp385b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107736.1 | NP_001101206.1 | 3 - 4 | 1613 | 69 | |
| XM_006234414.2 | XP_006234476.1 | 4 - 5 | 1272 | 69 | ||
| XM_006234416.2 | XP_006234478.1 | 3 - 4 | 1233 | 69 | ||
| XM_006234419.2 | XP_006234481.1 | 4 - 5 | 2429 | 69 | ||
| XM_006234420.2 | XP_006234482.1 | 4 - 5 | 798 | 69 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Zfp385b
Genomic Map