Assay Details
| Gene Symbol: Gpr155 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107811.1 | NP_001101281.1 | 12 - 13 | 2241 | 60 | |
| XM_006234354.2 | XP_006234416.1 | 13 - 14 | 2387 | 60 | ||
| XM_006234355.2 | XP_006234417.1 | 13 - 14 | 2383 | 60 | ||
| XM_006234356.2 | XP_006234418.1 | 13 - 14 | 2387 | 60 | ||
| XM_006234357.2 | XP_006234419.1 | 13 - 14 | 2387 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gpr155
Genomic Map