Assay Details
| Gene Symbol: Aven | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006234707.2 | XP_006234769.1 | 1 - 2 | 294 | 93 | |
| XM_008762093.1 | XP_008760315.1 | 1 - 2 | 294 | 93 | ||
More Information
Set Membership: |
|

Aven
Genomic Map