Assay Details
Gene Symbol: Ssrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031121.1 | NP_112383.1 | 12 - 13 | 1903 | 91 | |
XM_006234477.1 | XP_006234539.1 | 13 - 14 | 1907 | 91 | ||
XM_008761993.1 | XP_008760215.1 | 13 - 14 | 1889 | 91 | ||
XM_008761994.1 | XP_008760216.1 | 13 - 14 | 1813 | 91 | ||
GenBank mRNA | BC083588.1 | - | 12 - 13 | 1903 | 7132288 | 91 |
L08814.1 | - | 9 - 10 | 1166 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
