Assay Details
| Gene Symbol: Tas1r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271266.1 | NP_001258195.1 | 1 - 2 | 189 | 82 | |
| GenBank mRNA | AF127390.1 | - | 1 - 2 | 189 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tas1r2
Genomic Map