Assay Details
| Gene Symbol: Tas1r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271266.1 | NP_001258195.1 | 3 - 4 | 1269 | 62 | |
| GenBank mRNA | AF127390.1 | - | 3 - 4 | 1269 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tas1r2
Genomic Map