Assay Details
| Gene Symbol: Eif3i | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115035.1 | NP_001108507.1 | 42 | 55 | ||
| GenBank mRNA | BC158821.1 | - | 38 | 7375325 | 55 | |
| FQ230070.1 | - | 28 | 55 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the BC158821.1,NM_001115035.1 transcripts.
This assay is in the 5'UTR region of the BC158821.1,NM_001115035.1 transcripts.
Set Membership: |
|

Eif3i
Genomic Map