Assay Details
| Gene Symbol: Eif3i | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115035.1 | NP_001108507.1 | 138 | 63 | ||
| GenBank mRNA | BC158748.1 | - | 106 | 9101450 | 63 | |
| BC158821.1 | - | 134 | 7375325 | 63 | ||
| FQ209543.1 | - | 120 | 63 | |||
| FQ213040.1 | - | 119 | 63 | |||
| FQ220584.1 | - | 122 | 63 | |||
| FQ220807.1 | - | 123 | 63 | |||
| FQ227121.1 | - | 124 | 63 | |||
| FQ230070.1 | - | 124 | 63 | |||
| FQ231084.1 | - | 114 | 63 | |||
| FQ231314.1 | - | 114 | 63 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Eif3i
Genomic Map