Assay Details
| Gene Symbol: Eif3i | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115035.1 | NP_001108507.1 | 225 | 59 | ||
| GenBank mRNA | BC158748.1 | - | 193 | 9101450 | 59 | |
| BC158821.1 | - | 221 | 7375325 | 59 | ||
| FQ213040.1 | - | 206 | 59 | |||
| FQ220584.1 | - | 209 | 59 | |||
| FQ220807.1 | - | 210 | 59 | |||
| FQ227121.1 | - | 211 | 59 | |||
| FQ230070.1 | - | 211 | 59 | |||
| FQ231084.1 | - | 201 | 59 | |||
| FQ231314.1 | - | 201 | 59 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Eif3i
Genomic Map