Assay Details
| Gene Symbol: Eif3i | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115035.1 | NP_001108507.1 | 570 | 64 | ||
| GenBank mRNA | BC158748.1 | - | 538 | 9101450 | 64 | |
| BC158821.1 | - | 566 | 7375325 | 64 | ||
| FQ209543.1 | - | 552 | 64 | |||
| FQ213040.1 | - | 551 | 64 | |||
| FQ220584.1 | - | 554 | 64 | |||
| FQ220807.1 | - | 555 | 64 | |||
| FQ227121.1 | - | 556 | 64 | |||
| FQ230070.1 | - | 556 | 64 | |||
| FQ231084.1 | - | 546 | 64 | |||
| FQ231314.1 | - | 546 | 64 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Eif3i
Genomic Map