Assay Details
| Gene Symbol: Clcnkb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173103.1 | NP_775126.1 | 8 - 9 | 886 | 97 | |
| XM_006239320.2 | XP_006239382.1 | 11 - 12 | 3758 | 97 | ||
| XM_008764293.1 | XP_008762515.1 | 10 - 11 | 3426 | 97 | ||
| XM_008764294.1 | XP_008762516.1 | 10 - 11 | 1692 | 97 | ||
| XM_008764295.1 | XP_008762517.1 | 9 - 10 | 1837 | 97 | ||
| XM_008764296.1 | XP_008762518.1 | 9 - 10 | 1826 | 97 | ||
| XM_008764297.1 | XP_008762519.1 | 8 - 9 | 1697 | 97 | ||
| GenBank mRNA | BC081698.1 | - | 8 - 9 | 1381 | 97 | |
| D26111.1 | - | 9 - 10 | 940 | 97 | ||
| DQ437671.1 | - | 9 - 10 | 990 | 97 | ||
| DQ437672.1 | - | 9 - 10 | 883 | 97 | ||
| DQ437673.1 | - | 8 - 9 | 861 | 97 | ||
| DQ437674.1 | - | 8 - 9 | 825 | 97 | ||
| DQ437675.1 | - | 9 - 10 | 833 | 97 | ||
| DQ437676.1 | - | 8 - 9 | 811 | 97 | ||
| Z30663.1 | - | 8 - 9 | 886 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Clcnkb
Genomic Map