Assay Details
Gene Symbol: Utp11l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107978.2 | NP_001101448.1 | 6 - 7 | 637 | 79 | |
GenBank mRNA | BC168964.1 | - | 6 - 7 | 637 | 7130611 | 79 |
FQ226285.1 | - | 6 - 7 | 649 | 79 | ||
FQ231750.1 | - | 6 - 7 | 658 | 79 | ||
FQ233930.1 | - | 6 - 7 | 649 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
