Assay Details
| Gene Symbol: Adprhl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108680.1 | NP_001102150.1 | 3 - 4 | 569 | 83 | |
| GenBank mRNA | BC159416.1 | - | 3 - 4 | 539 | 8368851 | 83 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Adprhl2
Genomic Map