Assay Details
| Gene Symbol: Adprhl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108680.1 | NP_001102150.1 | 4 - 5 | 754 | 84 | |
| GenBank mRNA | BC159416.1 | - | 4 - 5 | 724 | 8368851 | 84 |
More Information
Updated Mapping Information
Set Membership: |
|

Adprhl2
Genomic Map