Assay Details
| Gene Symbol: Sh3d21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162535.1 | NP_001156007.1 | 6 - 7 | 495 | 102 | |
| XM_008764049.1 | XP_008762271.1 | 9 - 10 | 1654 | 102 | ||
| GenBank mRNA | BC162054.1 | - | 6 - 7 | 495 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sh3d21
Genomic Map