Assay Details
| Gene Symbol: Csf3r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106685.1 | NP_001100155.1 | 10 - 11 | 1465 | 67 | |
| XM_008764003.1 | XP_008762225.1 | 10 - 11 | 1736 | 67 | ||
| XM_008764004.1 | XP_008762226.1 | 9 - 10 | 1645 | 67 | ||
| XM_008764005.1 | XP_008762227.1 | 10 - 11 | 1564 | 67 | ||
| XM_008764006.1 | XP_008762228.1 | 9 - 10 | 1473 | 67 | ||
| XM_008764007.1 | XP_008762229.1 | 10 - 11 | 1703 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Csf3r
Genomic Map