Assay Details
| Gene Symbol: Pla2g2f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109587.1 | NP_001103057.1 | 1 - 2 | 235 | 71 | |
| XM_008764289.1 | XP_008762511.1 | 2 - 3 | 660 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pla2g2f
Genomic Map