Assay Details
| Gene Symbol: Gnl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025736.1 | NP_001020907.1 | 15 - 16 | 2119 | 93 | |
| GenBank mRNA | BC099173.1 | - | 15 - 16 | 2119 | 7382617 | 93 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P208741 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 424606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 266750 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534682 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 298204 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249926 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616257 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 355691 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 585054 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387084 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 638441 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 521570 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854499 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 176142 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939144 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 411124 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335631 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 290157 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 957136 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 739684 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872387 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151821 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 68277 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P208741 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P208741 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P208741 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P208741 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 60919 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 5622 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 35115 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 42615 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10871918 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871915 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871916 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871911 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871921 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871913 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871917 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871912 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871924 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871914 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871920 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871910 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871925 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871923 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871922 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871919 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10871909 | - | - |
| Rat Genome 230 2.0 Array | 1374804_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1384830_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1392010_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1392010_at | - | - |
| Rat Genome 230 2.0 Array | 1374804_at | - | - |
| Rat Genome 230 2.0 Array | 1384830_at | - | - |
| Affymetrix HT Rat Focus Array | 1374804_at | - | - |
| Affymetrix HT Rat Focus Array | 1392010_at | - | - |
| Affymetrix HT Rat Focus Array | 1392010_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1374804_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0500010111.rn.1 | TC0500001440.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500010112.rn.1 | TC0500001440.rn.1 | RTA TC |
| Rat Expression Set 230 | 1374804_at | - | - |
| Rat Expression Set 230 | 1374804_PM_at | - | - |
| Rat Expression Set 230 | 1392010_PM_at | - | - |
| Rat Expression Set 230 | 1392010_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871909 | - | - |
| Rat Genome U34 Set | rc_AI060050_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20731431 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1358784 | - | - |

Gnl2
Genomic Map