Assay Details
| Gene Symbol: Aunip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001071529.3 | XP_001071529.1 | 2 - 3 | 451 | 66 | |
| XM_006239131.2 | XP_006239193.1 | 2 - 3 | 571 | 66 | ||
More Information
Set Membership: |
|

Aunip
Genomic Map