Assay Details
| Gene Symbol: Uts2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019160.2 | NP_062033.1 | 2 - 3 | 70 | ||
| GenBank mRNA | AF172174.1 | - | 2 - 3 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Uts2
Genomic Map