Assay Details
| Gene Symbol: Phc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013169.2 | NP_001013187.2 | 11 - 12 | 2029 | 70 | |
| XM_006238926.2 | XP_006238988.1 | 12 - 13 | 2100 | 70 | ||
| XM_006238930.2 | XP_006238992.1 | 4 - 5 | 747 | 70 | ||
| XM_008764139.1 | XP_008762361.1 | 4 - 5 | 712 | 70 | ||
| GenBank mRNA | BC090339.1 | - | 1 - 2 | 93 | 70 | |
| BC169102.1 | - | 4 - 5 | 691 | 70 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC090339.1 transcripts.
Set Membership: |
|

Phc2
Genomic Map