Assay Details
| Gene Symbol: Trappc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008376.1 | NP_001008377.1 | 1 - 2 | 109 | 55 | |
| GenBank mRNA | BC086377.1 | - | 1 - 2 | 109 | 7301917 | 55 |
| FQ214189.1 | - | 1 - 2 | 165 | 55 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trappc3
Genomic Map