Assay Details
Gene Symbol: Arid1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106635.1 | NP_001100105.1 | 13 - 14 | 2582 | 62 | |
XM_006239011.2 | XP_006239073.1 | 14 - 15 | 3779 | 62 | ||
XM_006239012.2 | XP_006239074.1 | 14 - 15 | 3776 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
