Assay Details
| Gene Symbol: Igsf21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271454.1 | NP_001258383.1 | 3 - 4 | 874 | 61 | |
| XM_008764231.1 | XP_008762453.1 | 2 - 3 | 188 | 61 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the XM_008764231.1 transcripts.
Set Membership: |
|

Igsf21
Genomic Map