Assay Details
Gene Symbol: Plekhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127566.1 | NP_001121038.1 | 4 - 5 | 610 | 60 | |
GenBank mRNA | BC161896.1 | - | 4 - 5 | 610 | 7319876 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
