Assay Details
| Gene Symbol: Serpina1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022519.2 | NP_071964.2 | 3 - 4 | 879 | 88 | |
| XM_006240456.1 | XP_006240518.1 | 2 - 3 | 736 | 88 | ||
| GenBank mRNA | BC078824.1 | - | 3 - 4 | 879 | 7131535 | 88 |
| D00675.1 | - | 2 - 3 | 682 | 88 | ||
| FQ209479.1 | - | 1 - 2 | 193 | 88 | ||
| FQ209494.1 | - | 1 - 2 | 193 | 88 | ||
| FQ209517.1 | - | 1 - 2 | 193 | 88 | ||
| FQ209677.1 | - | 1 - 2 | 193 | 88 | ||
| FQ209686.1 | - | 1 - 2 | 193 | 88 | ||
| FQ209779.1 | - | 1 - 2 | 193 | 88 | ||
| FQ209814.1 | - | 1 - 2 | 194 | 89 | ||
| FQ209897.1 | - | 1 - 2 | 193 | 88 | ||
| FQ209914.1 | - | 1 - 2 | 88 | |||
| FQ210068.1 | - | 1 - 2 | 193 | 88 | ||
| FQ210169.1 | - | 1 - 2 | 193 | 88 | ||
| FQ210655.1 | - | 2 - 3 | 672 | 88 | ||
| FQ210685.1 | - | 1 - 2 | 193 | 88 | ||
| FQ210775.1 | - | 1 - 2 | 193 | 88 | ||
| FQ210832.1 | - | 1 - 2 | 113 | 88 | ||
| FQ210989.1 | - | 1 - 2 | 193 | 88 | ||
| FQ210996.1 | - | 1 - 2 | 193 | 88 | ||
| FQ211012.1 | - | 1 - 2 | 193 | 88 | ||
| FQ211042.1 | - | 1 - 2 | 193 | 88 | ||
| FQ211203.1 | - | 1 - 2 | 193 | 88 | ||
| FQ211353.1 | - | 1 - 2 | 193 | 88 | ||
| FQ211455.1 | - | 1 - 2 | 59 | 88 | ||
| FQ218226.1 | - | 1 - 2 | 193 | 88 | ||
| FQ218241.1 | - | 1 - 2 | 193 | 88 | ||
| FQ218252.1 | - | 1 - 2 | 193 | 88 | ||
| FQ218261.1 | - | 1 - 2 | 193 | 88 | ||
| FQ218555.1 | - | 1 - 2 | 193 | 88 | ||
| FQ218567.1 | - | 1 - 2 | 193 | 88 | ||
| FQ218586.1 | - | 1 - 2 | 193 | 88 | ||
| FQ218654.1 | - | 1 - 2 | 193 | 88 | ||
| FQ218694.1 | - | 1 - 2 | 193 | 88 | ||
| FQ218710.1 | - | 1 - 2 | 192 | 88 | ||
| FQ218857.1 | - | 1 - 2 | 193 | 88 | ||
| FQ218947.1 | - | 1 - 2 | 193 | 88 | ||
| FQ218955.1 | - | 1 - 2 | 194 | 88 | ||
| FQ219224.1 | - | 1 - 2 | 193 | 88 | ||
| FQ219252.1 | - | 1 - 2 | 193 | 88 | ||
| FQ219268.1 | - | 1 - 2 | 193 | 88 | ||
| FQ219276.1 | - | 1 - 2 | 193 | 88 | ||
| FQ219303.1 | - | 1 - 2 | 193 | 88 | ||
| FQ219383.1 | - | 1 - 2 | 193 | 88 | ||
| FQ219441.1 | - | 1 - 2 | 193 | 88 | ||
| FQ219458.1 | - | 1 - 2 | 193 | 88 | ||
| FQ219621.1 | - | 1 - 2 | 193 | 88 | ||
| FQ219652.1 | - | 1 - 2 | 193 | 88 | ||
| FQ219732.1 | - | 1 - 2 | 193 | 88 | ||
| FQ219741.1 | - | 2 - 3 | 673 | 88 | ||
| FQ219765.1 | - | 1 - 2 | 193 | 88 | ||
| M32247.1 | - | 1 - 2 | 619 | 88 | ||
| X16273.1 | - | 1 - 2 | 65 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Serpina1
Genomic Map