Assay Details
| Gene Symbol: Slc10a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017047.1 | NP_058743.1 | 4 - 5 | 873 | 102 | |
| GenBank mRNA | BC091152.1 | - | 3 - 4 | 823 | 7370832 | 102 |
| M77479.1 | - | 4 - 5 | 873 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc10a1
Genomic Map