Assay Details
| Gene Symbol: Ifi27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203410.1 | NP_981955.1 | 1 - 2 | 48 | 71 | |
| GenBank mRNA | BC060548.1 | - | 1 - 2 | 48 | 6919535 | 71 |
| FQ219860.1 | - | 1 - 2 | 106 | 71 | ||
| FQ220098.1 | - | 1 - 2 | 117 | 71 | ||
| FQ226998.1 | - | 1 - 2 | 81 | 71 | ||
| FQ233730.1 | - | 1 - 2 | 82 | 71 | ||
| FQ233981.1 | - | 1 - 2 | 117 | 71 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC060548.1,NM_203410.1 transcripts.
Set Membership: |
|

Ifi27
Genomic Map