Assay Details
Gene Symbol: Rps6ka5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108048.1 | NP_001101518.1 | 8 - 9 | 1133 | 60 | |
XM_006240437.2 | XP_006240499.1 | 9 - 10 | 1316 | 60 | ||
XM_006240438.2 | XP_006240500.1 | 8 - 9 | 1244 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription, DNA-templated
protein phosphorylation
histone phosphorylation
positive regulation of histone phosphorylation
positive regulation of histone acetylation
intracellular signal transduction
histone H3-S10 phosphorylation
histone H3-S28 phosphorylation
histone H2A-S1 phosphorylation
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of NF-kappaB transcription factor activity
interleukin-1-mediated signaling pathway
protein kinase activity
protein serine/threonine kinase activity
ATP binding
regulation of transcription, DNA-templated
protein phosphorylation
histone phosphorylation
positive regulation of histone phosphorylation
positive regulation of histone acetylation
intracellular signal transduction
histone H3-S10 phosphorylation
histone H3-S28 phosphorylation
histone H2A-S1 phosphorylation
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of NF-kappaB transcription factor activity
interleukin-1-mediated signaling pathway
protein kinase activity
protein serine/threonine kinase activity
ATP binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P701779 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 836964 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763235 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 822137 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588400 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6986 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 941657 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 663076 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 39284 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 904627 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 238796 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600146 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374264 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622434 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624814 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 847224 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589086 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 72332 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 410516 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 156835 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230574 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 558135 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 780669 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449988 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 302787 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 710676 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 564808 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164276 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 956797 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 398327 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758258 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571533 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614136 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 301363 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294894 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 852007 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 842156 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724238 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 67435 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55824 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 182309 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 159275 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 436934 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 354715 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 639107 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936946 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680435 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434431 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 324216 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 620049 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 132830 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 390361 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14779 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364104 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448522 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621854 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928424 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288390 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816158 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419575 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218931 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374352 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 410133 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546346 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244555 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940185 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203855 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 885293 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21051 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 194812 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576400 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 961470 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744360 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 675984 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 751682 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272679 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901605 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453596 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 222555 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873126 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 1362 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 238178 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763087 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 718350 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 237314 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854876 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407805 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409875 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 192647 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 738049 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225970 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600522 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634339 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 490130 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837265 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 443442 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 165214 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 880302 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 307252 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 454810 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 937058 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908455 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 931821 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 463634 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810341 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118224 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 102033 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529148 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633137 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831915 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696205 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 350119 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741492 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363655 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569128 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 695666 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482961 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963564 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 467687 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 523854 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7910 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 85889 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16666 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 60962 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 158177 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 21162 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 143207 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 11400 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 127056 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 30453 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 85871 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 122171 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 93934 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 68329 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 2306 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 78646 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154312 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 28735 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33864 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 98710 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P701779 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P701779 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P701779 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P219402 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P21439 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 750 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 38854 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10891667 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891662 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891661 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891664 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891669 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891678 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891673 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891672 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891660 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891663 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891670 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891666 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891674 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891675 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891677 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891676 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891668 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10891659 | - | - |
Rat Genome 230 2.0 Array | 1398582_PM_at | - | - |
Rat Genome 230 2.0 Array | 1398582_at | - | - |
Affymetrix HT Rat Focus Array | 1382271_at | - | - |
Affymetrix HT Rat Focus Array | 1398582_at | - | - |
Affymetrix HT Rat Focus Array | 1398582_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1382271_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0600021333.rn.1 | TC0600003025.rn.1 | RTA TC |
Affymetrix Rat Gene 1.0 ST Array | 10891659 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20706579 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1371123 | - | - |
