Assay Details
| Gene Symbol: Cox16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163153.1 | NP_001156625.1 | 1 - 2 | 217 | 66 | |
| XM_006240298.2 | XP_006240360.1 | 1 - 2 | 251 | 66 | ||
| GenBank mRNA | FQ210472.1 | - | 1 - 2 | 210 | 66 | |
| FQ218255.1 | - | 1 - 2 | 199 | 66 | ||
| FQ224072.1 | - | 1 - 2 | 205 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cox16
Genomic Map