Assay Details
Gene Symbol: Lrrc74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037783.2 | NP_001032872.2 | 10 - 11 | 1554 | 60 | |
GenBank mRNA | BC083569.1 | - | 10 - 11 | 1503 | 60 | |
BC098927.1 | - | 1566 | 60 | |||
BC127444.1 | - | 10 - 11 | 1545 | 7135116 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P127753 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P157087 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31831 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273426 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838834 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915661 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 165593 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804340 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633341 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230981 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 238477 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 85943 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 33644 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636801 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698411 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731107 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 646172 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 530897 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127272 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736313 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947808 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673991 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 200139 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102436 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75327 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 35241 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P127753 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P157087 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P157087 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P127753 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P127753 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P157087 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P157087 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 91966 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 28113 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 103290 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 22018 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10886143 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10886144 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10886149 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10886145 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10886141 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10886152 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10886142 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10886155 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10886147 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10886148 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10886153 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10886156 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10886151 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10886154 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10886146 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10886150 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10886140 | - | - |
Affymetrix Rat Clariom S Assay | PSR0600008671.rn.1 | TC0600001216.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0600008649.rn.1 | TC0600001216.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0600008652.rn.1 | TC0600001216.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0600008653.rn.1 | TC0600001216.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0600008659.rn.1 | TC0600001216.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0600008676.rn.1 | TC0600001216.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0600008648.rn.1 | TC0600001216.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0600008651.rn.1 | TC0600001216.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0600008669.rn.1 | TC0600001216.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0600008674.rn.1 | TC0600001216.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0600008677.rn.1 | TC0600001216.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0600008650.rn.1 | TC0600001216.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0600008660.rn.1 | TC0600001216.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0600008672.rn.1 | TC0600001216.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0600008654.rn.1 | TC0600001216.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0600008667.rn.1 | TC0600001216.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0600008668.rn.1 | TC0600001216.rn.1 | RTA TC |
Affymetrix Rat Gene 1.0 ST Array | 10886140 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21267359 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1372895 | - | - |
