Assay Details
| Gene Symbol: Dcaf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108716.1 | NP_001102186.1 | 5 - 6 | 560 | 63 | |
| XM_006240347.2 | XP_006240409.1 | 5 - 6 | 560 | 63 | ||
| XM_008764814.1 | XP_008763036.1 | 4 - 5 | 608 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dcaf4
Genomic Map